Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008598.2 | NP_032624.1 | 3 - 4 | 373 | 61 | |
GenBank mRNA | AB092489.1 | - | 2 - 3 | 284 | 61 | |
AB092490.1 | - | 2 - 3 | 284 | 61 | ||
AK090389.1 | - | 363 | 61 | |||
BC031888.1 | - | 3 - 4 | 342 | 5372377 | 61 | |
BC096557.1 | - | 3 - 4 | 376 | 1366095 | 61 | |
M84524.1 | - | 2 - 3 | 299 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |