Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198029.2 | NP_932146.2 | 11 - 12 | 2102 | 69 | |
GenBank mRNA | AF073489.1 | - | 8 - 9 | 888 | 69 | |
AK030947.1 | - | 10 - 11 | 2292 | 69 | ||
AK050804.1 | - | 10 - 11 | 1250 | 69 | ||
BC029093.1 | - | 2 - 3 | 221 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |