Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146119.1 | NP_001139591.1 | 4 - 5 | 445 | 97 | |
NM_029758.4 | NP_084034.2 | 5 - 6 | 518 | 97 | ||
GenBank mRNA | AK047142.1 | - | 4 - 5 | 408 | 97 | |
BC049664.1 | - | 5 - 6 | 480 | 6532813 | 97 | |
BC052465.1 | - | 4 - 5 | 426 | 6834768 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |