Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269481 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269479 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269480 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269482 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2053491 |
BC006865.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2053491 |
AK167386.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2053491 |
AK168136.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2053491 |
AK168279.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2053491 |
AK152960.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
52091 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
320682 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
682004 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
AK031344.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
AK078469.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
AK138747.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
BC023130.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
BC040763.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
BC084730.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
BC099670.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
BC099693.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
NM_009177.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914024 |
X73523.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
399238 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
26536 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
481385 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
755759 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
848326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
280702 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
436956 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
719123 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45149 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
107634 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153997 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P121697 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P174800 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2053491 |
BC006865.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2053491 |
AK167386.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2053491 |
AK168136.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2053491 |
AK168279.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2053491 |
AK152960.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269490 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269488 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269463 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269485 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269487 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269459 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269465 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269489 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269495 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269470 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269460 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269468 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269472 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269497 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269474 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269462 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269484 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269492 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269479 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269493 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269498 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269478 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269477 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269482 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28269480 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02470936 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P121697 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P174800 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P135455 |
AK148928.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P70810 |
BC006865.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P70810 |
AK167386.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P70810 |
AK168136.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P70810 |
AK168279.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P70810 |
AK152960.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P339495 |
BC006865.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P339495 |
AK167386.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P339495 |
AK168136.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P339495 |
AK168279.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P339495 |
AK152960.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
AK148928.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
AK151008.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
AK076558.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P297968 |
NM_027959.3 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P174800 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P121697 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10334237 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10334238 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09893679 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10334242 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09893681 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269480 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269481 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269479 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28269482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269564 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269477 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269543 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269580 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269562 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269602 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28269500 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09940578 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10136524 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09851733 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269513 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269541 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269507 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09851732 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269525 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269470 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269474 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269556 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269511 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269501 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269547 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09805951 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10246066 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269517 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269465 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269500 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269521 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269453 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269529 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269527 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09893680 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269479 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10334242 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269490 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269457 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269478 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269555 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269498 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269463 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269502 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269518 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09851726 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269553 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269524 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269454 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269523 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10278898 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269503 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269466 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269484 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269493 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269542 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269468 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10334241 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269495 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269510 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269487 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269492 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09895493 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269509 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269559 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269506 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269548 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269557 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28269460 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09893667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09895496 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09893674 |
- |
- |