Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130452.1 | NP_569719.1 | 1 - 2 | 305 | 114 | |
GenBank mRNA | AY033514.1 | - | 1 - 2 | 243 | 114 | |
BC019406.1 | - | 1 - 2 | 305 | 5134018 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |