Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026613.4 | NP_080889.3 | 1 - 2 | 402 | 79 | |
GenBank mRNA | AK012383.1 | - | 1 - 2 | 373 | 79 | |
AK014021.1 | - | 1 - 2 | 255 | 79 | ||
AK019172.1 | - | 1 - 2 | 384 | 79 | ||
AK019247.1 | - | 1 - 2 | 373 | 79 | ||
AK033555.1 | - | 1 - 2 | 381 | 79 | ||
AK147056.1 | - | 1 - 2 | 403 | 79 | ||
BC049229.1 | - | 1 - 2 | 135 | 79 | ||
BC062248.1 | - | 1 - 2 | 348 | 79 | ||
BC089458.1 | - | 1 - 2 | 239 | 79 | ||
BC108339.1 | - | 1 - 2 | 385 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |