Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290830.1 | NP_001277759.1 | 17 - 18 | 3783 | 93 | |
NM_013735.4 | NP_038763.3 | 17 - 18 | 3783 | 93 | ||
GenBank mRNA | AJ414734.1 | - | 16 - 17 | 3626 | 93 | |
BC035206.1 | - | 5 - 6 | 995 | 3979746 | 93 | |
BC079906.1 | - | 17 - 18 | 3717 | 30639375 | 93 | |
CA315900.1 | - | 1 - 2 | 454 | 93 | ||
CA316961.1 | - | 1 - 2 | 458 | 93 | ||
U67885.1 | - | 1 - 2 | 129 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |