Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700038731.mm.1 |
NM_001161624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038731.mm.1 |
NM_001161624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038731.mm.1 |
NM_009876.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038731.mm.1 |
NM_009876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038731.mm.1 |
TC0700004585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038730.mm.1 |
NM_001161624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038730.mm.1 |
NM_001161624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038730.mm.1 |
NM_009876.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038730.mm.1 |
NM_009876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038730.mm.1 |
TC0700004585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038734.mm.1 |
NM_001161624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038734.mm.1 |
NM_001161624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038734.mm.1 |
NM_009876.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038734.mm.1 |
NM_009876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038734.mm.1 |
TC0700004585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038735.mm.1 |
NM_001161624.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038735.mm.1 |
NM_001161624.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038735.mm.1 |
NM_009876.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038735.mm.1 |
NM_009876.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038735.mm.1 |
TC0700004585.mm.1 |
MTA TC |