Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009358.3 | NP_033384.2 | 4 - 5 | 643 | 94 | |
GenBank mRNA | AB053464.1 | - | 4 - 5 | 588 | 94 | |
AK154050.1 | - | 4 - 5 | 619 | 94 | ||
AK160587.1 | - | 4 - 5 | 584 | 94 | ||
AY338234.1 | - | 4 - 5 | 573 | 94 | ||
BC010716.1 | - | 4 - 5 | 556 | 2655861 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |