Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207204.2 | NP_997087.2 | 16 - 17 | 2401 | 69 | |
GenBank mRNA | BC067071.1 | - | 16 - 17 | 2335 | 30615484 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |