Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207204.2 | NP_997087.2 | 9 - 10 | 1349 | 76 | |
GenBank mRNA | AK129253.1 | - | 9 - 10 | 1282 | 76 | |
BC067071.1 | - | 9 - 10 | 1283 | 30615484 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |