Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK145279.1 | - | 2 - 3 | 350 | 73 | |
BC024588.1 | - | 2 - 3 | 221 | 5066294 | 73 | |
BY735425.1 | - | 2 - 3 | 350 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |