Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253885.1 | NP_001240814.1 | 11 - 12 | 1962 | 64 | |
NM_001253886.1 | NP_001240815.1 | 10 - 11 | 1956 | 64 | ||
NM_001253887.1 | NP_001240816.1 | 11 - 12 | 962 | 64 | ||
NM_009685.3 | NP_033815.1 | 10 - 11 | 1770 | 64 | ||
GenBank mRNA | AK030748.1 | - | 10 - 11 | 1653 | 64 | |
AK075730.1 | - | 11 - 12 | 942 | 64 | ||
AK083830.1 | - | 11 - 12 | 1692 | 64 | ||
AK144340.1 | - | 11 - 12 | 1963 | 64 | ||
AK164140.1 | - | 11 - 12 | 1687 | 64 | ||
BC022732.1 | - | 4 - 5 | 337 | 64 | ||
BC036956.1 | - | 10 - 11 | 1600 | 64 | ||
BC048395.1 | - | 9 - 10 | 1108 | 6392016 | 64 | |
L77865.1 | - | 1 - 2 | 94 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |