Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111111.1 | NP_001104581.1 | 12 - 13 | 1318 | 72 | |
GenBank mRNA | AB476647.1 | - | 11 - 12 | 1194 | 72 | |
AB476648.1 | - | 12 - 13 | 1257 | 72 | ||
AK010773.1 | - | 11 - 12 | 1239 | 72 | ||
AK131109.1 | - | 1088 | 72 | |||
AK155125.1 | - | 12 - 13 | 1308 | 72 | ||
AK157410.1 | - | 12 - 13 | 1297 | 72 | ||
AK158186.1 | - | 11 - 12 | 1432 | 72 | ||
BC060054.1 | - | 11 - 12 | 1655 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |