Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111111.1 | NP_001104581.1 | 5 - 6 | 719 | 83 | |
GenBank mRNA | AB476647.1 | - | 5 - 6 | 658 | 83 | |
AB476648.1 | - | 5 - 6 | 658 | 83 | ||
AK155125.1 | - | 5 - 6 | 709 | 83 | ||
AK157410.1 | - | 5 - 6 | 698 | 83 | ||
AK158186.1 | - | 5 - 6 | 83 | |||
BC060054.1 | - | 5 - 6 | 882 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |