Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141931.1 | NP_001135403.1 | 1 - 2 | 326 | 129 | |
NM_001141932.1 | NP_001135404.1 | 1 - 2 | 326 | 129 | ||
GenBank mRNA | AK145897.1 | - | 1 - 2 | 328 | 129 | |
AK161102.1 | - | 1 - 2 | 276 | 129 | ||
AK164307.1 | - | 1 - 2 | 320 | 129 | ||
BC057866.1 | - | 1 - 2 | 358 | 5327143 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |