Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177833.1 | NP_001171304.1 | 3 - 4 | 638 | 73 | |
NM_001177834.1 | NP_001171305.1 | 3 - 4 | 638 | 73 | ||
NM_001177835.1 | NP_001171306.1 | 3 - 4 | 638 | 73 | ||
NM_001177836.1 | NP_001171307.1 | 3 - 4 | 638 | 73 | ||
NM_001177837.1 | NP_001171308.1 | 3 - 4 | 638 | 73 | ||
NM_001177838.1 | NP_001171309.1 | 3 - 4 | 638 | 73 | ||
NM_001177839.1 | NP_001171310.1 | 3 - 4 | 638 | 73 | ||
NM_145533.2 | NP_663508.1 | 3 - 4 | 638 | 73 | ||
GenBank mRNA | AF495851.1 | - | 3 - 4 | 506 | 73 | |
AF495852.1 | - | 3 - 4 | 507 | 73 | ||
AF495853.1 | - | 3 - 4 | 507 | 73 | ||
AJ567473.1 | - | 3 - 4 | 557 | 73 | ||
AJ567474.1 | - | 3 - 4 | 557 | 73 | ||
AJ567476.1 | - | 3 - 4 | 557 | 73 | ||
AJ567477.1 | - | 3 - 4 | 557 | 73 | ||
AJ567478.1 | - | 3 - 4 | 557 | 73 | ||
AJ567479.1 | - | 3 - 4 | 557 | 73 | ||
AJ567480.1 | - | 3 - 4 | 557 | 73 | ||
AK143129.1 | - | 3 - 4 | 612 | 73 | ||
AK159275.1 | - | 3 - 4 | 565 | 73 | ||
AK159372.1 | - | 3 - 4 | 565 | 73 | ||
AK166350.1 | - | 537 | 73 | |||
BC004831.1 | - | 3 - 4 | 557 | 2647695 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |