Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008027.2 | NP_032053.1 | 11 - 12 | 1248 | 86 | |
GenBank mRNA | AF145044.1 | - | 10 - 11 | 1089 | 86 | |
AK150692.1 | - | 11 - 12 | 1270 | 86 | ||
AK150827.1 | - | 11 - 12 | 1197 | 86 | ||
AK167323.1 | - | 11 - 12 | 1175 | 86 | ||
AK190295.1 | - | 1 - 2 | 102 | 86 | ||
AY167925.1 | - | 11 - 12 | 1254 | 86 | ||
BC004647.1 | - | 11 - 12 | 1258 | 3158071 | 86 | |
U90435.1 | - | 11 - 12 | 1248 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |