Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008027.2 | NP_032053.1 | 1 - 2 | 144 | 95 | |
GenBank mRNA | AK150692.1 | - | 1 - 2 | 166 | 95 | |
AK150827.1 | - | 1 - 2 | 93 | 95 | ||
AK167323.1 | - | 1 - 2 | 71 | 95 | ||
AY167925.1 | - | 1 - 2 | 150 | 95 | ||
BC004647.1 | - | 1 - 2 | 154 | 3158071 | 95 | |
U90435.1 | - | 1 - 2 | 144 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |