Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK044224.1 | - | 5 - 6 | 443 | 70 | |
AK083340.1 | - | 5 - 6 | 465 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |