Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023059.3 | NP_075546.2 | 5 - 6 | 1076 | 72 | |
GenBank mRNA | AF239957.1 | - | 4 - 5 | 587 | 72 | |
AK133518.1 | - | 5 - 6 | 701 | 72 | ||
AK145889.1 | - | 5 - 6 | 1079 | 72 | ||
BC010806.1 | - | 4 - 5 | 598 | 4240425 | 72 | |
BC094069.1 | - | 5 - 6 | 682 | 4974962 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |