Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282072.1 | NP_001269001.1 | 1 - 2 | 454 | 75 | |
NM_001282076.1 | NP_001269005.1 | 2 - 3 | 411 | 75 | ||
NM_016874.3 | NP_058570.1 | 1 - 2 | 454 | 75 | ||
GenBank mRNA | AF102818.1 | - | 1 - 2 | 314 | 75 | |
AK082115.1 | - | 1 - 2 | 307 | 75 | ||
AK144333.1 | - | 1 - 2 | 223 | 75 | ||
AK146546.1 | - | 1 - 2 | 331 | 75 | ||
AK155836.1 | - | 1 - 2 | 420 | 75 | ||
FJ377318.1 | - | 2 - 3 | 271 | 75 | ||
FJ377319.1 | - | 1 - 2 | 299 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |