Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081268.1 | NP_001074737.1 | 8 - 9 | 1331 | 90 | |
GenBank mRNA | AK220244.1 | - | 8 - 9 | 1288 | 90 | |
BC039632.1 | - | 5 - 6 | 901 | 90 | ||
BC147509.1 | - | 8 - 9 | 1328 | 9056457 | 90 | |
BC151010.1 | - | 8 - 9 | 1328 | 9087923 | 90 | |
BC151017.1 | - | 8 - 9 | 1328 | 9087930 | 90 | |
BC172059.1 | - | 8 - 9 | 1328 | 9054345 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |