Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033319.2 | NP_001028491.1 | 22 - 23 | 3102 | 97 | |
NM_001195086.1 | NP_001182015.1 | 23 - 24 | 3177 | 97 | ||
GenBank mRNA | BC157936.1 | - | 23 - 24 | 2994 | 9088116 | 97 |
BC158007.1 | - | 22 - 23 | 3059 | 9007315 | 97 | |
BC172096.1 | - | 23 - 24 | 2997 | 9054382 | 97 | |
EU568869.1 | - | 23 - 24 | 3177 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |