Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199188.1 | NP_001186117.1 | 8 - 9 | 952 | 74 | |
NM_024225.5 | NP_077187.1 | 7 - 8 | 863 | 74 | ||
GenBank mRNA | AK007676.1 | - | 7 - 8 | 811 | 74 | |
AK029463.1 | - | 7 - 8 | 823 | 74 | ||
AK031873.1 | - | 8 - 9 | 844 | 74 | ||
AK049129.1 | - | 8 - 9 | 844 | 74 | ||
AK078776.1 | - | 8 - 9 | 845 | 74 | ||
AK144710.1 | - | 8 - 9 | 848 | 74 | ||
AK152311.1 | - | 8 - 9 | 889 | 74 | ||
AK159115.1 | - | 4 - 5 | 406 | 74 | ||
AK159904.1 | - | 7 - 8 | 768 | 74 | ||
AK167362.1 | - | 7 - 8 | 783 | 74 | ||
AK169545.1 | - | 6 - 7 | 1758 | 74 | ||
AK169786.1 | - | 8 - 9 | 844 | 74 | ||
AK212372.1 | - | 2 - 3 | 178 | 74 | ||
BC002242.1 | - | 8 - 9 | 921 | 3492262 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |