Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012038.4 | NP_036168.1 | 1 - 2 | 292 | 141 | |
GenBank mRNA | AJ489331.1 | - | 1 - 2 | 256 | 141 | |
AK144454.1 | - | 1 - 2 | 222 | 141 | ||
AK160224.1 | - | 1 - 2 | 224 | 141 | ||
BC046226.1 | - | 1 - 2 | 197 | 5692284 | 141 | |
BY264558.1 | - | 1 - 2 | 298 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |