Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029744.1 | - | 17 - 18 | 2607 | 81 | |
AK030138.1 | - | 17 - 18 | 2562 | 81 | ||
DQ867028.1 | - | 17 - 18 | 2607 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |