Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178778.3 | NP_848893.2 | 15 - 16 | 1481 | 67 | |
GenBank mRNA | AK044772.1 | - | 15 - 16 | 1477 | 67 | |
AK082408.1 | - | 16 - 17 | 1583 | 67 | ||
BC132485.1 | - | 15 - 16 | 1456 | 40130762 | 67 | |
BC144986.1 | - | 16 - 17 | 1557 | 67 | ||
BC144987.1 | - | 15 - 16 | 1532 | 67 | ||
BC144989.1 | - | 15 - 16 | 1455 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |