Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178890.3 | NP_849221.2 | 12 - 13 | 2917 | 95 | |
GenBank mRNA | AK042322.1 | - | 12 - 13 | 2917 | 95 | |
AK151620.1 | - | 12 - 13 | 2913 | 95 | ||
AK152163.1 | - | 12 - 13 | 2913 | 95 | ||
AK170569.1 | - | 12 - 13 | 2916 | 95 | ||
AK172414.1 | - | 12 - 13 | 2915 | 95 | ||
AK172468.1 | - | 12 - 13 | 2914 | 95 | ||
BC006756.1 | - | 3 - 4 | 457 | 95 | ||
BC054399.1 | - | 12 - 13 | 2917 | 6493840 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |