Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178890.3 | NP_849221.2 | 16 - 17 | 3297 | 99 | |
GenBank mRNA | AK042322.1 | - | 16 - 17 | 3297 | 99 | |
AK151620.1 | - | 16 - 17 | 3293 | 99 | ||
AK152163.1 | - | 16 - 17 | 3293 | 99 | ||
AK170569.1 | - | 16 - 17 | 3294 | 99 | ||
AK172414.1 | - | 16 - 17 | 3295 | 99 | ||
AK172468.1 | - | 16 - 17 | 3294 | 99 | ||
BC006756.1 | - | 7 - 8 | 837 | 99 | ||
BC054399.1 | - | 16 - 17 | 3297 | 6493840 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |