Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173749.4 | NP_776110.3 | 4 - 5 | 657 | 132 | |
GenBank mRNA | AK031254.1 | - | 4 - 5 | 529 | 132 | |
AK088017.1 | - | 4 - 5 | 556 | 132 | ||
BB620582.1 | - | 4 - 5 | 524 | 132 | ||
BC031841.1 | - | 4 - 5 | 611 | 3675412 | 132 | |
BC057685.1 | - | 4 - 5 | 611 | 3675412 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |