Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200012722.mm.1 |
NM_026620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012722.mm.1 |
NM_026620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012722.mm.1 |
TC0200001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012726.mm.1 |
NM_026620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012726.mm.1 |
NM_026620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012726.mm.1 |
TC0200001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012732.mm.1 |
NM_026620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012732.mm.1 |
NM_026620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012732.mm.1 |
TC0200001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012729.mm.1 |
NM_026620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012729.mm.1 |
NM_026620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012729.mm.1 |
TC0200001724.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012730.mm.1 |
NM_026620.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012730.mm.1 |
NM_026620.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012730.mm.1 |
TC0200001724.mm.1 |
MTA TC |