Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045842.1 | 1 - 2 | 161 | 140 | ||
GenBank mRNA | AK016851.1 | - | 1 - 2 | 161 | 140 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KCNQ1 overlapping transcript 1 | Kcnq1ot1 |
Kvlqt1-as, Lit1, Tssc8 |
-33.2kb | Sense | Chr. 7 - 143213111 - 143296547 on NCBI Build GRCm38 |
Downstream | cyclin-dependent kinase inhibitor 1C (P57) | Cdkn1c |
AL024410, CDKI, Kip2, p57(kip2), p57Kip2 |
127.7kb | Sense | Chr. 7 - 143458339 - 143461072 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |