Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173449.3 | NP_775625.1 | 1 - 2 | 277 | 84 | |
GenBank mRNA | AK076579.1 | - | 1 - 2 | 278 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mus musculus Probe spans exons |