Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026115.4 | NP_080391.2 | 9 - 10 | 1001 | 78 | |
GenBank mRNA | AK010715.1 | - | 9 - 10 | 1011 | 78 | |
AK014330.1 | - | 9 - 10 | 1012 | 78 | ||
AK018270.1 | - | 9 - 10 | 992 | 78 | ||
AK041700.1 | - | 9 - 10 | 1005 | 78 | ||
AK169315.1 | - | 9 - 10 | 1004 | 78 | ||
BC038262.1 | - | 7 - 8 | 881 | 78 | ||
BC055460.1 | - | 8 - 9 | 968 | 4457298 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |