Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081329.1 | NP_001074798.1 | 3 - 4 | 1141 | 61 | |
GenBank mRNA | AK034954.1 | - | 3 - 4 | 1143 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |