Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205214.1 | NP_001192143.1 | 2 - 3 | 631 | 55 | |
NM_001205215.1 | NP_001192144.1 | 2 - 3 | 354 | 55 | ||
NM_001205216.1 | NP_001192145.1 | 2 - 3 | 354 | 55 | ||
NM_011306.4 | NP_035436.1 | 2 - 3 | 631 | 55 | ||
GenBank mRNA | AK140371.1 | - | 2 - 3 | 370 | 55 | |
AK159671.1 | - | 2 - 3 | 632 | 55 | ||
BC019432.1 | - | 2 - 3 | 286 | 5132301 | 55 | |
BC049773.1 | - | 2 - 3 | 591 | 6581828 | 55 | |
KJ650219.1 | - | 2 - 3 | 445 | 55 | ||
KJ663717.1 | - | 2 - 3 | 445 | 55 | ||
KJ663718.1 | - | 3 - 4 | 331 | 55 | ||
KJ663719.1 | - | 3 - 4 | 331 | 55 | ||
KJ663720.1 | - | 2 - 3 | 262 | 55 | ||
KJ663721.1 | - | 3 - 4 | 316 | 55 | ||
M26804.1 | - | 1 - 2 | 223 | 55 | ||
M84818.1 | - | 2 - 3 | 304 | 55 | ||
X66224.1 | - | 1 - 2 | 223 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |