Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011581.3 | NP_035711.2 | 8 - 9 | 1696 | 66 | |
GenBank mRNA | AK132508.1 | - | 8 - 9 | 1527 | 66 | |
AK142552.1 | - | 8 - 9 | 1670 | 66 | ||
BC053702.1 | - | 8 - 9 | 1498 | 30062369 | 66 | |
L07803.1 | - | 8 - 9 | 1524 | 66 | ||
M64866.1 | - | 8 - 9 | 1524 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |