Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110794.1 | NP_001104264.1 | 8 - 9 | 975 | 152 | |
NM_009674.3 | NP_033804.2 | 8 - 9 | 1090 | 152 | ||
GenBank mRNA | AK032013.1 | - | 8 - 9 | 866 | 152 | |
AK077579.1 | - | 8 - 9 | 783 | 152 | ||
AK134589.1 | - | 9 - 10 | 860 | 152 | ||
AK149654.1 | - | 8 - 9 | 782 | 152 | ||
AK151438.1 | - | 8 - 9 | 784 | 152 | ||
AK161613.1 | - | 8 - 9 | 784 | 152 | ||
AK167588.1 | - | 8 - 9 | 758 | 152 | ||
AK169199.1 | - | 8 - 9 | 757 | 152 | ||
BC008997.1 | - | 8 - 9 | 774 | 3157926 | 152 | |
L13129.1 | - | 8 - 9 | 777 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |