Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146515.2 | NP_666726.1 | 1 | 361 | 79 | |
GenBank mRNA | BC106777.1 | - | 1 | 361 | 40040271 | 79 |
BC106778.1 | - | 1 | 361 | 40040272 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon |