Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfnb59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080711.2 | NP_001074180.1 | 4 - 5 | 813 | 93 | |
GenBank mRNA | DQ365828.1 | - | 4 - 5 | 813 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |