Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011195.2 | NP_035325.1 | 2 - 3 | 496 | 77 | |
GenBank mRNA | BC119062.1 | - | 2 - 3 | 421 | 8733811 | 77 |
BC120746.1 | - | 2 - 3 | 421 | 40129669 | 77 | |
U16958.1 | - | 2 - 3 | 496 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |