Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025611.5 | NP_079887.3 | 24 - 25 | 4921 | 92 | |
GenBank mRNA | AK010907.1 | - | 3 - 4 | 446 | 92 | |
AK147156.1 | - | 24 - 25 | 4679 | 92 | ||
AK147905.1 | - | 24 - 25 | 4641 | 92 | ||
AK170808.1 | - | 24 - 25 | 4694 | 92 | ||
AK172882.1 | - | 23 - 24 | 3994 | 92 | ||
BC019645.1 | - | 14 - 15 | 2209 | 92 | ||
BC024345.1 | - | 3 - 4 | 713 | 92 | ||
BC026946.1 | - | 6 - 7 | 1029 | 92 | ||
BC059865.1 | - | 20 - 21 | 3434 | 92 | ||
BC065096.1 | - | 23 - 24 | 4408 | 92 | ||
BC100544.1 | - | 3 - 4 | 557 | 92 | ||
BC138998.1 | - | 24 - 25 | 4859 | 8862020 | 92 | |
BC139000.1 | - | 24 - 25 | 4859 | 8862022 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |