Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008300.3 | NP_032326.3 | 10 - 11 | 1556 | 152 | |
GenBank mRNA | AK053601.1 | - | 10 - 11 | 1490 | 152 | |
AK054211.1 | - | 3 - 4 | 278 | 152 | ||
AK133772.1 | - | 10 - 11 | 1498 | 152 | ||
AK155305.1 | - | 10 - 11 | 1473 | 152 | ||
AK166102.1 | - | 10 - 11 | 1541 | 152 | ||
AK220167.1 | - | 10 - 11 | 1507 | 152 | ||
BC003770.1 | - | 10 - 11 | 1465 | 3494648 | 152 | |
BC003950.1 | - | 10 - 11 | 1451 | 152 | ||
CN458707.1 | - | 4 - 5 | 498 | 152 | ||
D85904.1 | - | 10 - 11 | 1258 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |