Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026173.3 | NP_080449.1 | 7 - 8 | 1027 | 68 | |
GenBank mRNA | AK004769.2 | - | 7 - 8 | 955 | 68 | |
AK012926.1 | - | 7 - 8 | 995 | 68 | ||
AK098149.1 | - | 7 - 8 | 920 | 68 | ||
AK150656.1 | - | 7 - 8 | 1028 | 68 | ||
BC016527.1 | - | 7 - 8 | 941 | 3157281 | 68 | |
BC027118.1 | - | 7 - 8 | 965 | 5359153 | 68 | |
BC037631.1 | - | 7 - 8 | 984 | 4954391 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |