Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankdd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042714.1 | NP_001036179.1 | 12 - 13 | 1411 | 69 | |
GenBank mRNA | AK033386.1 | - | 6 - 7 | 651 | 69 | |
AK033960.1 | - | 5 - 6 | 605 | 69 | ||
BC112417.1 | - | 12 - 13 | 1408 | 40061167 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |