Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC016231.1 | - | 16 - 17 | 1323 | 4488798 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |