Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022011.4 | NP_071294.3 | 6 - 7 | 496 | 145 | |
GenBank mRNA | AB221674.1 | - | 5 - 6 | 310 | 145 | |
AK078297.1 | - | 6 - 7 | 467 | 145 | ||
BC016231.1 | - | 6 - 7 | 451 | 4488798 | 145 | |
BC053382.1 | - | 6 - 7 | 460 | 30047823 | 145 | |
DV662169.1 | - | 4 - 5 | 236 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |