Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030026.2 | NP_084302.1 | 1 - 2 | 257 | 86 | |
GenBank mRNA | AK132265.1 | - | 1 - 2 | 237 | 86 | |
AK143233.1 | - | 1 - 2 | 258 | 86 | ||
AK145564.1 | - | 1 - 2 | 209 | 86 | ||
AK146844.1 | - | 1 - 2 | 217 | 86 | ||
AK146865.1 | - | 1 - 2 | 207 | 86 | ||
BC027640.1 | - | 1 - 2 | 192 | 86 | ||
BC141193.1 | - | 1 - 2 | 223 | 9055805 | 86 | |
BC141194.1 | - | 1 - 2 | 223 | 9055806 | 86 | |
BY138007.1 | - | 1 - 2 | 247 | 86 | ||
BY237319.1 | - | 1 - 2 | 258 | 86 | ||
BY276243.1 | - | 1 - 2 | 239 | 86 | ||
BY713615.1 | - | 1 - 2 | 237 | 86 | ||
CK620826.1 | - | 1 - 2 | 212 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |