Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aggf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025630.3 | NP_079906.2 | 5 - 6 | 1169 | 114 | |
GenBank mRNA | AY500995.1 | - | 5 - 6 | 1142 | 114 | |
BC052410.1 | - | 5 - 6 | 1146 | 5716440 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |